accessibility menu, dialog, popup

Test

CAS RN 93919-56-3

CAS RN 93919-56-3

IUPAC Name: [4-(trifluoromethoxy)phenyl]methanaminium
Synonyms: [4-(trifluoromethoxy)phenyl]methanaminium
Molecular Weight (g/mol): 192.16
Molecular Formula: C8H9F3NO
InChi Key: DBGROTRFYBSUTR-UHFFFAOYSA-O
SMILES: [NH3+]CC1=CC=C(OC(F)(F)F)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)