CAS RN 939-23-1
CAS RN 939-23-1
IUPAC Name:
4-phenylpyridine
Synonyms:
4-phenylpyridine azabiphenyl
Molecular Weight (g/mol):
155.2
Molecular Formula:
C11H9N
InChi Key:
JVZRCNQLWOELDU-UHFFFAOYSA-N
SMILES:
C1=CC=C(C=C1)C1=CC=NC=C1
No Refinements
1
–
0
of
0
results
1
4-Phenylpyridine, 99%
CAS: 939-23-1 Molecular Formula: C11H9N Molecular Weight (g/mol): 155.20 MDL Number: MFCD00006420 InChI Key: JVZRCNQLWOELDU-UHFFFAOYSA-N Synonym: pyridine, 4-phenyl,p-phenylpyridine,4-phenyl-pyridine,4-azabiphenyl,unii-452kd9ycg7,4-aza-1,1'-biphenyl,phenyl-pyridine,4-phenylpyridin,4-pyridylbenzene,zlchem 480 PubChem CID: 13651 IUPAC Name: 4-phenylpyridine SMILES: C1=CC=C(C=C1)C1=CC=NC=C1