CAS RN 938-96-5
CAS RN 938-96-5
IUPAC Name:
(2S)-2-(4-hydroxyphenyl)propanoate
Synonyms:
(2S)-2-(4-hydroxyphenyl)propanoate
Molecular Weight (g/mol):
165.17
Molecular Formula:
C9H9O3
InChi Key:
ZHMMPVANGNPCBW-LURJTMIESA-M
SMILES:
C[C@H](C([O-])=O)C1=CC=C(O)C=C1
1
–
2
of
2
results
1
2-(4-Hydroxyphenyl)propionic acid, 98%, Thermo Scientific Chemicals
CAS: 938-96-5 Molecular Formula: C9H10O3 Molecular Weight (g/mol): 166.176 MDL Number: MFCD00128847 InChI Key: ZHMMPVANGNPCBW-UHFFFAOYSA-N Synonym: 2-4-hydroxyphenyl propanoic acid,2-4-hydroxyphenyl propionic acid,4-hydroxyhydratropate,4-hydroxyhydratropic acid,4-hydroxyphenyl-2-propionic acid,2-4'-hydroxyphenyl-propionic acid,2-4-hydroxyphenyl propionate,acmc-209ro0,4-hydroxyphenyl propanoic acid,methyl 4-hydroxyphenylacetic acid PubChem CID: 102526 ChEBI: CHEBI:1868 IUPAC Name: 2-(4-hydroxyphenyl)propanoic acid SMILES: CC(C1=CC=C(C=C1)O)C(=O)O