CAS RN 936643-80-0
CAS RN 936643-80-0
IUPAC Name:
2-(chloromethyl)pyrimidine hydrochloride
Synonyms:
2-(chloromethyl)pyrimidine hydrochloride
Molecular Weight (g/mol):
165.02
Molecular Formula:
C5H6Cl2N2
InChi Key:
XLGVMJXAZRCTRU-UHFFFAOYSA-N
SMILES:
Cl.ClCC1=NC=CC=N1