accessibility menu, dialog, popup

UserName

CAS RN 936-59-4

ClO

CAS RN 936-59-4

IUPAC Name: 3-chloro-1-phenylpropan-1-one
Synonyms: 3-chloro-1-phenylpropan-1-one
Molecular Weight (g/mol): 168.62
Molecular Formula: C9H9ClO
InChi Key: KTJRGPZVSKWRTJ-UHFFFAOYSA-N
SMILES: ClCCC(=O)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results