accessibility menu, dialog, popup

UserName

CAS RN 935-30-8

OHN

CAS RN 935-30-8

IUPAC Name: azonan-2-one
Synonyms: 2-azacyclo-nonanone
Molecular Weight (g/mol): 141.21
Molecular Formula: C8H15NO
InChi Key: YDLSUFFXJYEVHW-UHFFFAOYSA-N
SMILES: O=C1CCCCCCCN1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)