accessibility menu, dialog, popup

UserName

CAS RN 93465-26-0

CH3OO

CAS RN 93465-26-0

IUPAC Name: 2'-methoxy-[1,1'-biphenyl]-2-carbaldehyde
Synonyms: 2'-methoxy-[1,1'-biphenyl]-2-carbaldehyde
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: ZYNNXRUYVVQLOW-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C1=CC=CC=C1C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results