CAS RN 934570-59-9
CAS RN 934570-59-9
IUPAC Name:
{1-[(pyridin-3-yl)methyl]piperidin-4-yl}methanol
Synonyms:
[1-(pyridin-3-ylmethyl)piperidin-4-yl]methanol
Molecular Weight (g/mol):
206.29
Molecular Formula:
C12H18N2O
InChi Key:
RAJQONMXWKPTTO-UHFFFAOYSA-N
SMILES:
OCC1CCN(CC2=CN=CC=C2)CC1
1
–
1
of
1
results
1
[1-(Pyrid-3-ylmethyl)piperid-4-yl]methanol, 97%, Thermo Scientific™
CAS: 934570-59-9 Molecular Formula: C12H18N2O Molecular Weight (g/mol): 206.289 MDL Number: MFCD09879988 InChI Key: RAJQONMXWKPTTO-UHFFFAOYSA-N Synonym: 1-pyrid-3-ylmethyl piperid-4-yl methanol,1-pyridin-3-ylmethyl piperidin-4-yl methanol,4-piperidinemethanol,1-3-pyridinylmethyl,1-3-pyridylmethyl-4-piperidyl methan-1-ol,4-piperidinemethanol, 1-3-pyridinylmethyl-, dihydrochloride PubChem CID: 24229775 IUPAC Name: [1-(pyridin-3-ylmethyl)piperidin-4-yl]methanol SMILES: C1CN(CCC1CO)CC2=CN=CC=C2