accessibility menu, dialog, popup

UserName

CAS RN 934570-55-5

CH3NNHOCH3

CAS RN 934570-55-5

IUPAC Name: [4-(3,5-dimethyl-1H-pyrazol-1-yl)phenyl]methanol
Synonyms: [4-(3,5-dimethylpyrazol-1-yl)phenyl]methanol
Molecular Weight (g/mol): 202.26
Molecular Formula: C12H14N2O
InChi Key: HROWBOXYWTWPCS-UHFFFAOYSA-N
SMILES: CC1=NN(C(C)=C1)C1=CC=C(CO)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results