accessibility menu, dialog, popup

Test

CAS RN 930111-03-8

CAS RN 930111-03-8

IUPAC Name: {3-[2-(chloromethyl)phenoxy]propyl}dimethylamine hydrochloride
Synonyms: {3-[2-(chloromethyl)phenoxy]propyl}dimethylamine hydrochloride
Molecular Weight (g/mol): 264.19
Molecular Formula: C12H19Cl2NO
InChi Key: FJINIPMBLHIDJV-UHFFFAOYSA-N
SMILES: Cl.CN(C)CCCOC1=CC=CC=C1CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results