accessibility menu, dialog, popup

UserName

BrSO

CAS RN 930-96-1

IUPAC Name: 3-bromothiophene-2-carbaldehyde
Synonyms: 3-bromothiophene-2-carbaldehyde
Molecular Weight (g/mol): 191.04
Molecular Formula: C5H3BrOS
InChi Key: BCZHCWCOQDRYGS-UHFFFAOYSA-N
SMILES: BrC1=C(SC=C1)C=O
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (3)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)