accessibility menu, dialog, popup

UserName

CAS RN 92954-90-0

(S)OHHOHNOOO

CAS RN 92954-90-0

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-hydroxyphenyl)propanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid
Molecular Weight (g/mol): 403.43
Molecular Formula: C24H21NO5
InChi Key: SWZCTMTWRHEBIN-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=C(O)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results