accessibility menu, dialog, popup

UserName

CH3NNH3CH3C

CAS RN 92507-97-6

IUPAC Name: 1-ethyl-2,3-dimethyl-1H-imidazol-3-ium
Synonyms: 3-ethyl-1,2-dimethylimidazol-1-ium
Molecular Weight (g/mol): 125.19
Molecular Formula: C7H13N2
InChi Key: IRGDPGYNHSIIJJ-UHFFFAOYSA-N
SMILES: CCN1C=C[N+](C)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results