accessibility menu, dialog, popup

UserName

CAS RN 92-29-5

NN(E/Z)(E/Z)

CAS RN 92-29-5

IUPAC Name: {phenyl[(phenylmethylidene)amino]methyl}(phenylmethylidene)amine
Synonyms: {phenyl[(phenylmethylidene)amino]methyl}(phenylmethylidene)amine
Molecular Weight (g/mol): 298.39
Molecular Formula: C21H18N2
InChi Key: VUYRFIIFTJICNA-UHFFFAOYSA-N
SMILES: C(=NC(N=CC1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results