accessibility menu, dialog, popup

UserName

CAS RN 917-64-6

H3CMgI

CAS RN 917-64-6

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 166.24
Molecular Formula: CH3IMg
InChi Key: VXWPONVCMVLXBW-UHFFFAOYSA-M
SMILES: [CH3-].[Mg++].[I-]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results