accessibility menu, dialog, popup

UserName

H3COFFFOHO

CAS RN 916421-04-0

IUPAC Name: 2-[3-methoxy-5-(trifluoromethyl)phenyl]acetic acid
Synonyms: [3-methoxy-5-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 234.17
Molecular Formula: C10H9F3O3
InChi Key: QROADCOZBAMFID-UHFFFAOYSA-N
SMILES: COC1=CC(CC(O)=O)=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results