accessibility menu, dialog, popup

UserName

CAS RN 916420-84-3

H3CONH2FFF

CAS RN 916420-84-3

IUPAC Name: 1-[4-methoxy-2-(trifluoromethyl)phenyl]methanamine
Synonyms: 1-[4-methoxy-2-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol): 205.18
Molecular Formula: C9H10F3NO
InChi Key: GRRJRPNJFJEEPK-UHFFFAOYSA-N
SMILES: COC1=CC(=C(CN)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results