accessibility menu, dialog, popup

UserName

ClOFCl

CAS RN 916420-61-6

IUPAC Name: 3,6-dichloro-2-fluorobenzaldehyde
Synonyms: 3,6-dichloro-2-fluorobenzaldehyde
Molecular Weight (g/mol): 193
Molecular Formula: C7H3Cl2FO
InChi Key: ZYBCMWVCTUCCQP-UHFFFAOYSA-N
SMILES: FC1=C(Cl)C=CC(Cl)=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results