accessibility menu, dialog, popup

UserName

CAS RN 916420-57-0

CH3OFFFOOH

CAS RN 916420-57-0

IUPAC Name: 3-[3-methyl-5-(trifluoromethoxy)phenyl]propanoic acid
Synonyms: 3-[3-methyl-5-(trifluoromethoxy)phenyl]propanoic acid
Molecular Weight (g/mol): 248.2
Molecular Formula: C11H11F3O3
InChi Key: LEZTWHDBDSJSTN-UHFFFAOYSA-N
SMILES: CC1=CC(CCC(O)=O)=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results