accessibility menu, dialog, popup

UserName

CAS RN 914635-52-2

IUPAC Name: 3-bromo-5-(trifluoromethoxy)benzonitrile
Synonyms: 3-bromo-5-(trifluoromethoxy)benzonitrile
Molecular Weight (g/mol): 266.02
Molecular Formula: C8H3BrF3NO
InChi Key: PQPKERBGCBHRJO-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC(Br)=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results