accessibility menu, dialog, popup

UserName

CAS RN 913835-79-7

IUPAC Name: [3-(hydrazinecarbonyl)phenyl]boronic acid
Synonyms: 3-(hydrazinecarbonyl)phenylboronic acid
Molecular Weight (g/mol): 179.97
Molecular Formula: C7H9BN2O3
InChi Key: NKQNSAXAGLKZBP-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results