accessibility menu, dialog, popup

UserName

CAS RN 913835-37-7

BOHNCH3ClHOOH

CAS RN 913835-37-7

IUPAC Name: {4-[(3-chloro-4-methylphenyl)carbamoyl]phenyl}boronic acid
Synonyms: 4-[(3-chloro-4-methylphenyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 289.52
Molecular Formula: C14H13BClNO3
InChi Key: IZMAVYDCXUROFV-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=C(NC(=O)C2=CC=C(C=C2)B(O)O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results