CAS RN 912288-64-3
CAS RN 912288-64-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
YHO-13351 free base, MedChemExpress
MedChemExpress YHO-13351 free base is the prodrug of YHO-13177, which is a potent and specific inhibitor of BCRP.