CAS RN 910463-68-2
CAS RN 910463-68-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Semaglutide, MedChemExpress
MedChemExpress Semaglutide, a long-acting GLP-1 analogue, is a glucagon-like peptide-1 (GLP-1) receptor agonist. Semaglutide has the potential for type 2 diabetes treatment.