accessibility menu, dialog, popup

UserName

CAS RN 90719-29-2

H3CON(S)OOCH3CH3(E)

CAS RN 90719-29-2

IUPAC Name: (4S)-3-[(2E)-but-2-enoyl]-4-(propan-2-yl)-1,3-oxazolidin-2-one
Synonyms: (4S)-3-[(2E)-but-2-enoyl]-4-isopropyl-1,3-oxazolidin-2-one
Molecular Weight (g/mol): 197.23
Molecular Formula: C10H15NO3
InChi Key: YZRHIMNEVFYTAJ-WTSVBCDHSA-N
SMILES: C\C=C\C(=O)N1[C@H](COC1=O)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results