CAS RN 906650-60-0
CAS RN 906650-60-0
IUPAC Name:
1-(2-phenylethyl)-4-(2-phenylethynyl)benzene
Synonyms:
1-(2-phenylethyl)-4-(2-phenylethynyl)benzene
Molecular Weight (g/mol):
282.39
Molecular Formula:
C22H18
InChi Key:
GXNVEXGZHSEHNG-UHFFFAOYSA-N
SMILES:
C(CC1=CC=C(C=C1)C#CC1=CC=CC=C1)C1=CC=CC=C1