accessibility menu, dialog, popup

Test

CAS RN 906352-66-7

CAS RN 906352-66-7

IUPAC Name: methyl({[2-(thiomorpholin-4-yl)pyridin-4-yl]methyl})amine
Synonyms: methyl({[2-(thiomorpholin-4-yl)pyridin-4-yl]methyl})amine
Molecular Weight (g/mol): 223.34
Molecular Formula: C11H17N3S
InChi Key: KEXCZGSQJYTNRX-UHFFFAOYSA-N
SMILES: CNCC1=CC(=NC=C1)N1CCSCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results