accessibility menu, dialog, popup

UserName

CAS RN 904696-62-4

NNNH2HCl

CAS RN 904696-62-4

IUPAC Name: 1-{4-[(1H-pyrazol-1-yl)methyl]phenyl}methanamine hydrochloride
Synonyms: 1-[4-(pyrazol-1-ylmethyl)phenyl]methanamine hydrochloride
Molecular Weight (g/mol): 223.7
Molecular Formula: C11H14ClN3
InChi Key: QUVMWCVEYSYIOW-UHFFFAOYSA-N
SMILES: Cl.NCC1=CC=C(CN2C=CC=N2)C=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results