accessibility menu, dialog, popup

UserName

CAS RN 902837-35-8

IUPAC Name: 3-[2-(propan-2-yl)phenoxy]piperidine
Synonyms: 3-(2-isopropylphenoxy)piperidine
Molecular Weight (g/mol): 219.33
Molecular Formula: C14H21NO
InChi Key: DFJMRKMGDKDNAX-UHFFFAOYNA-N
SMILES: CC(C)C1=CC=CC=C1OC1CCCNC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results