accessibility menu, dialog, popup

UserName

CAS RN 902836-16-2

H3CHNONNNH

CAS RN 902836-16-2

IUPAC Name: N-ethyl-2-(piperazin-1-yl)pyridine-3-carboxamide
Synonyms: N-ethyl-2-(piperazin-1-yl)pyridine-3-carboxamide
Molecular Weight (g/mol): 234.3
Molecular Formula: C12H18N4O
InChi Key: JYFVWJXFJQURPD-UHFFFAOYSA-N
SMILES: CCNC(=O)C1=C(N=CC=C1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results