accessibility menu, dialog, popup

UserName

CAS RN 892501-89-2

H3CNHNS

CAS RN 892501-89-2

IUPAC Name: methyl({[3-(1,3-thiazol-2-yl)phenyl]methyl})amine
Synonyms: methyl({[3-(1,3-thiazol-2-yl)phenyl]methyl})amine
Molecular Weight (g/mol): 204.29
Molecular Formula: C11H12N2S
InChi Key: MNTPWZTYKCKBQX-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC(=C1)C1=NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results