accessibility menu, dialog, popup

UserName

CAS RN 88912-26-9

NClClOOH

CAS RN 88912-26-9

IUPAC Name: 2,5-dichloropyridine-4-carboxylic acid
Synonyms: 2,5-dichloropyridine-4-carboxylic acid
Molecular Weight (g/mol): 192
Molecular Formula: C6H3Cl2NO2
InChi Key: GFOVTTQVBDEYPP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Cl)=NC=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)