accessibility menu, dialog, popup

UserName

CAS RN 888711-52-2

IUPAC Name: potassium [(cyclohexylamino)methyl]trifluoroboranuide
Synonyms: potassium [(cyclohexylamino)methyl]trifluoroboranuide
Molecular Weight (g/mol): 219.1
Molecular Formula: C7H14BF3KN
InChi Key: GYUWPHIATXXXJT-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)CNC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results