accessibility menu, dialog, popup

UserName

CAS RN 886536-37-4

BOFFFHOOH

CAS RN 886536-37-4

IUPAC Name: [4-(2,2,2-trifluoroethoxy)phenyl]boronic acid
Synonyms: 4-(2,2,2-trifluoroethoxy)phenylboronic acid
Molecular Weight (g/mol): 219.95
Molecular Formula: C8H8BF3O3
InChi Key: DWRAMTYYMNLVBD-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(OCC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results