accessibility menu, dialog, popup

UserName

CAS RN 886501-99-1

FFFBrClCl

CAS RN 886501-99-1

IUPAC Name: 2-(bromomethyl)-3,4-dichloro-1-(trifluoromethyl)benzene
Synonyms: 2-(bromomethyl)-3,4-dichloro-1-(trifluoromethyl)benzene
Molecular Weight (g/mol): 307.92
Molecular Formula: C8H4BrCl2F3
InChi Key: BTXOBZONYMZJCP-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(CBr)C(Cl)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results