accessibility menu, dialog, popup

UserName

H3COFFOOH

CAS RN 886501-88-8

IUPAC Name: 2-(2,3-difluoro-6-methoxyphenyl)acetic acid
Synonyms: (2,3-difluoro-6-methoxyphenyl)acetic acid
Molecular Weight (g/mol): 202.16
Molecular Formula: C9H8F2O3
InChi Key: SPSVPCOXZRUNQI-UHFFFAOYSA-N
SMILES: COC1=C(CC(O)=O)C(F)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results