accessibility menu, dialog, popup

UserName

CAS RN 886501-83-3

H3COFFBr

CAS RN 886501-83-3

IUPAC Name: 2-(bromomethyl)-3,4-difluoro-1-methoxybenzene
Synonyms: 2-(bromomethyl)-3,4-difluoro-1-methoxybenzene
Molecular Weight (g/mol): 237.04
Molecular Formula: C8H7BrF2O
InChi Key: SVUCRCMGEHPJGQ-UHFFFAOYSA-N
SMILES: COC1=C(CBr)C(F)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results