accessibility menu, dialog, popup

UserName

OOHOFFFF

CAS RN 886501-32-2

IUPAC Name: 2-[4-fluoro-3-(trifluoromethoxy)phenyl]acetic acid
Synonyms: [4-fluoro-3-(trifluoromethoxy)phenyl]acetic acid
Molecular Weight (g/mol): 238.14
Molecular Formula: C9H6F4O3
InChi Key: RICXEJYYIREWPA-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(OC(F)(F)F)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results