accessibility menu, dialog, popup

UserName

CAS RN 886501-20-8

NH2OFFFF

CAS RN 886501-20-8

IUPAC Name: 1-[4-fluoro-3-(trifluoromethoxy)phenyl]methanamine
Synonyms: 1-[4-fluoro-3-(trifluoromethoxy)phenyl]methanamine
Molecular Weight (g/mol): 209.14
Molecular Formula: C8H7F4NO
InChi Key: LQEZTMITGPWONC-UHFFFAOYSA-N
SMILES: NCC1=CC(OC(F)(F)F)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results