accessibility menu, dialog, popup

UserName

CAS RN 886498-89-1

OHOFOFFF

CAS RN 886498-89-1

IUPAC Name: 3-fluoro-4-(trifluoromethoxy)benzoic acid
Synonyms: 3-fluoro-4-(trifluoromethoxy)benzoic acid
Molecular Weight (g/mol): 224.11
Molecular Formula: C8H4F4O3
InChi Key: PIBFTHWKVKHHMA-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(F)=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results