accessibility menu, dialog, popup

UserName

CAS RN 886498-55-1

H3COFBrF

CAS RN 886498-55-1

IUPAC Name: 2-(bromomethyl)-1,3-difluoro-4-methoxybenzene
Synonyms: 2-(bromomethyl)-1,3-difluoro-4-methoxybenzene
Molecular Weight (g/mol): 237.04
Molecular Formula: C8H7BrF2O
InChi Key: JCGVLYZGZNSCQD-UHFFFAOYSA-N
SMILES: COC1=C(F)C(CBr)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results