accessibility menu, dialog, popup

UserName

CH3OFFFCl

CAS RN 886497-07-0

IUPAC Name: 1-chloro-3-methoxy-5-(trifluoromethyl)benzene
Synonyms: 1-chloro-3-methoxy-5-(trifluoromethyl)benzene
Molecular Weight (g/mol): 210.58
Molecular Formula: C8H6ClF3O
InChi Key: RZLMBMVNHVUJML-UHFFFAOYSA-N
SMILES: COC1=CC(Cl)=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results