CAS RN 886496-99-7
IUPAC Name:
2-[3-chloro-5-(trifluoromethyl)phenyl]acetic acid
Synonyms:
[3-chloro-5-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol):
238.59
Molecular Formula:
C9H6ClF3O2
InChi Key:
VEEZRFPBIRDYOE-UHFFFAOYSA-N
SMILES:
OC(=O)CC1=CC(=CC(Cl)=C1)C(F)(F)F
1
–
1
of
1
results
1
3-Chloro-5-(trifluoromethyl)phenylacetic acid, 97%, Thermo Scientific™
CAS: 886496-99-7 Molecular Formula: C9H6ClF3O2 Molecular Weight (g/mol): 238.59 MDL Number: MFCD04115851 InChI Key: VEEZRFPBIRDYOE-UHFFFAOYSA-N Synonym: 2-3-chloro-5-trifluoromethyl phenyl acetic acid,3-chloro-5-trifluoromethyl phenylacetic acid,3-chloro-5-trifluorophenylacetic acid,3'-chloro-5'-trifluoromethylbenzeneacetic acid,3-chloro-5-trifluoromethyl phenyl acetic acid,3-chloro-5-trifluoromethyl benzeneacetic acid,pubchem24111,3-chloro-5-trifluoromethylphenylacetic acid,2-5-chloro-3-trifluoromethyl phenyl acetic acid PubChem CID: 3797627 IUPAC Name: 2-[3-chloro-5-(trifluoromethyl)phenyl]acetic acid SMILES: OC(=O)CC1=CC(=CC(Cl)=C1)C(F)(F)F