accessibility menu, dialog, popup

UserName

CAS RN 885267-96-9

FFFSOOCl

CAS RN 885267-96-9

IUPAC Name: 3'-(trifluoromethyl)-[1,1'-biphenyl]-4-sulfonyl chloride
Synonyms: 3'-(trifluoromethyl)-[1,1'-biphenyl]-4-sulfonyl chloride
Molecular Weight (g/mol): 320.71
Molecular Formula: C13H8ClF3O2S
InChi Key: WJSSWLPZDVPLIK-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(=C1)C1=CC=C(C=C1)S(Cl)(=O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)