CAS RN 885-11-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Nitrosoantipyrine, TRC
CAS: 885-11-0 Molecular Formula: C11 H11 N3 O2 Molecular Weight (g/mol): 217.22 Synonym: 3H-Pyrazol-3-one, 1,2-dihydro-1,5-dimethyl-4-nitroso-2-phenyl- (9CI, ACI),1,2-Dihydro-1,5-dimethyl-4-nitroso-2-phenyl-3H-pyrazol-3-one (ACI),Antipyrine, 4-nitroso- (6CI, 7CI, 8CI),1,2-Dihydro-2,3-dimethyl-4-nitroso-1-phenylpyrazol-5-one,1,5-Dimethyl-4-nitroso-2-phenyl-2,3-dihydro-1H-pyrazol-3-one,1-Phenyl-2,3-dimethyl-4-nitroso-5-pyrazolone,2,3-Dimethyl-4-nitroso-1-phenyl-5-pyrazolone,4-Nitrosoantipyrin,4-Nitrosoantipyrine,Nitrosoantipyrine,NSC 163932,NSC 75060 IUPAC Name: 1,5-dimethyl-4-nitroso-2-phenylpyrazol-3-one SMILES: CN1N(C(=O)C(=C1C)N=O)c2ccccc2