accessibility menu, dialog, popup

UserName

CAS RN 883502-07-6

FFBrOHO

CAS RN 883502-07-6

IUPAC Name: 2-(2-bromo-4,5-difluorophenyl)acetic acid
Synonyms: (2-bromo-4,5-difluorophenyl)acetic acid
Molecular Weight (g/mol): 251.03
Molecular Formula: C8H5BrF2O2
InChi Key: FQSLXVRCDYUESO-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(F)=C(F)C=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results