accessibility menu, dialog, popup

UserName

CAS RN 875664-40-7

OFFFNOH

CAS RN 875664-40-7

IUPAC Name: 2-hydroxy-5-(trifluoromethoxy)benzonitrile
Synonyms: 2-hydroxy-5-(trifluoromethoxy)benzonitrile
Molecular Weight (g/mol): 203.12
Molecular Formula: C8H4F3NO2
InChi Key: GOAVRYAOBXORSY-UHFFFAOYSA-N
SMILES: OC1=C(C=C(OC(F)(F)F)C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results