CAS RN 874101-00-5
CAS RN 874101-00-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
RO4987655, MedChemExpress
MedChemExpress RO4987655 is an orally active and highly selective MEK inhibitor with an IC50 of 5.2 nM for inhibition of MEK1/MEK2.