accessibility menu, dialog, popup

UserName

CAS RN 871125-83-6

BOOCH3CH3H3CCH3NCl

CAS RN 871125-83-6

IUPAC Name: 7-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Synonyms: 7-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Molecular Weight (g/mol): 289.57
Molecular Formula: C15H17BClNO2
InChi Key: GZDUYRAAWRRVEY-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=NC2=CC(Cl)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results