accessibility menu, dialog, popup

UserName

CAS RN 870482-12-5

IUPAC Name: O-[11-(trimethoxysilyl)undecyl]hydroxylamine
Synonyms: O-[11-(trimethoxysilyl)undecyl]hydroxylamine
Molecular Weight (g/mol): 307.51
Molecular Formula: C14H33NO4Si
InChi Key: SDAAYIFGIWPAIZ-UHFFFAOYSA-N
SMILES: CO[Si](CCCCCCCCCCCON)(OC)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results